Methyl-4'-chloro-3'-fluoro[1,1'-biphenyl]-4-carboxylate ≥95%

Supplier: Matrix Scientific

046040-500MG 046040-1G 046040-5G
101873-866EA 375.38 USD
101873-866 101873-868 101873-870
Methyl-4'-chloro-3'-fluoro[1,1'-biphenyl]-4-carboxylate ≥95%
Methyl-4'-chloro-3'-fluoro[1,1'-biphenyl]-4-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₀ClFO₂
MW: 264.69 g/mol
Melting Pt: 88…90 °C
MDL Number: MFCD06801758
CAS Number: 886362-12-5

Order Now

Specification Test Results

Melting point 88-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR