Methyl-3',4'-dichloro[1,1'-biphenyl]-4-carboxylate ≥95%

Supplier: Matrix Scientific

046038-500MG 046038-1G 046038-5G
101873-854EA 375.38 USD
101873-854 101873-856 101873-858
Methyl-3',4'-dichloro[1,1'-biphenyl]-4-carboxylate ≥95%
Methyl-3',4'-dichloro[1,1'-biphenyl]-4-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₀Cl₂O₂
MW: 281.14 g/mol
Melting Pt: 115…117 °C
MDL Number: MFCD08276964
CAS Number: 886362-11-4

Order Now

Specification Test Results

Melting point 115-117°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR