Ethyl-2-aminonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-aminonicotinate

046036-5G 046036-25G
101873-846EA 78.46 USD
101873-846 101873-848
Ethyl-2-aminonicotinate ≥95%
Ethyl 2-aminonicotinate

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: 96-98°C
MDL Number: MFCD01569452
CAS Number: 13362-26-0

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR