Methyl-4-chlorobenzo[b]thiophene-2-carboxylate ≥95%
Supplier: Matrix Scientific
046034-500MG
046034-1G
046034-5G
101873-834EA
181.54
USD
101873-834
101873-836
101873-838
Methyl-4-chlorobenzo[b]thiophene-2-carboxylate ≥95%
Methyl 4-chlorobenzo[b]thiophene-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₇ClO₂S Melting Pt: 95-97°C |
MDL Number:
MFCD07371546 CAS Number: 35212-95-4 |
Specification Test Results
Melting point | 95-97°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...