2-(4-Benzylpiperidino)-1,3-thiazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

046031-500MG 046031-1G
101873-818EA 1296.92 USD
101873-818 101873-820
2-(4-Benzylpiperidino)-1,3-thiazole-5-carbaldehyde ≥95%
2-(4-Benzylpiperidino)-1,3-thiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₁₆H₁₈N₂OS
MW: 286.4 g/mol
Melting Pt: 98…100 °C
MDL Number: MFCD08276935
CAS Number: 886362-09-0

Order Now

Specification Test Results

Melting point 98-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR