2-(tert-Butyl)-1H-indole ≥95%

Supplier: Matrix Scientific

046028-500MG 046028-1G
101873-810EA 556.92 USD
101873-810 101873-812
2-(tert-Butyl)-1H-indole ≥95%
2-(tert-Butyl)-1H-indole

Hazard: Irritant

Formula: C₁₂H₁₅N
Melting Pt: 75…76 °C
MDL Number: MFCD04004060
CAS Number: 1805-65-8

Order Now

Specification Test Results

Melting point 75-76°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR