tert-Butyl-4-hydroxydihydro-2(3H)-isoxazolecarboxylate ≥95%

Supplier: Matrix Scientific

046026-500MG 046026-1G 046026-5G
101873-800EA 921.54 USD
101873-800 101873-802 101873-804
tert-Butyl-4-hydroxydihydro-2(3H)-isoxazolecarboxylate ≥95%
tert-Butyl-4-hydroxydihydro-2(3H)-isoxazolecarboxylate

Hazard: Irritant

Formula: C₈H₁₅NO₄
MW: 189.21 g/mol
Melting Pt: 35…37 °C
MDL Number: MFCD07698596
CAS Number: 262444-52-0

Order Now

Specification Test Results

Melting point 35-37°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR