2-Acetyl-4,5-dibromothiophene ≥95%
Supplier: Matrix Scientific
046025-500MG
046025-1G
046025-5G
101873-794EA
375.38
USD
101873-794
101873-796
101873-798
2-Acetyl-4,5-dibromothiophene ≥95%
2-Acetyl-4,5-dibromothiophene
Hazard: Irritant
Formula:
C₆H₄Br₂OS Melting Pt: 85…86 °C |
CAS Number:
7209-12-3 |
Specification Test Results
Melting point | 83-85°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...