N-{3-[3-(1-Pyrrolidinyl)-1-propynyl]phenyl}acetamide ≥95%

Supplier: Matrix Scientific

046022-500MG 046022-1G
101873-786EA 556.92 USD
101873-786 101873-788
N-{3-[3-(1-Pyrrolidinyl)-1-propynyl]phenyl}acetamide ≥95%
N-{3-[3-(1-Pyrrolidinyl)-1-propynyl]phenyl}acetamide

Hazard: Irritant

Formula: C₁₅H₁₈N₂O
MW: 242.32 g/mol
Melting Pt: 127…128 °C
MDL Number: MFCD06795473
CAS Number: 885950-23-2

Order Now

Specification Test Results

Melting point 127-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR