N-[3-(3-Morpholino-1-propynyl)phenyl]acetamide ≥95%

Supplier: Matrix Scientific

046021-500MG 046021-1G 046021-5G
101873-780EA 556.92 USD
101873-780 101873-782 101873-784
N-[3-(3-Morpholino-1-propynyl)phenyl]acetamide ≥95%
N-[3-(3-Morpholino-1-propynyl)phenyl]acetamide

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₂
Melting Pt: 110…112 °C
MDL Number: MFCD06795474
CAS Number: 885950-22-1

Order Now

Specification Test Results

Melting point 110-112°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR