2-[(4-Bromophenyl)methylene]malononitrile ≥95%

Supplier: Matrix Scientific

Synonyms: (4-Bromobenzylidene)malononitrile, 2-(4-Bromobenzylidene)malononitrile

046020-1G 046020-5G
101873-776EA 232.31 USD
101873-776 101873-778
2-[(4-Bromophenyl)methylene]malononitrile ≥95%
2-[(4-Bromophenyl)methylene]malononitrile

Hazard: Irritant

Formula: C₁₀H₅BrN₂
MW: 233.07 g/mol
Melting Pt: 160…162 °C
MDL Number: MFCD00176213
CAS Number: 2826-24-6

Order Now

Specification Test Results

Melting point 160-162°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR