2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole ≥95%

Supplier: Matrix Scientific

046019-500MG 046019-1G 046019-5G
101873-768EA 140 USD
101873-768 101873-770 101873-772
2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole ≥95%
2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole

Hazard: Irritant

Formula: C₉H₇ClN₂O
Melting Pt: 119…121 °C
MDL Number: MFCD00466332
CAS Number: 33575-83-6

Order Now

Specification Test Results

Melting point 119-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR