2-Acetyl-3,4-dibromothiophene ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(3,4-Dibromothiophen-2-yl)ethanone, 1-(3,4-Dibromothien-2-yl)ethanone
046018-500MG
046018-1G
046018-5G
101873-762EA
375.38
USD
101873-762
101873-764
101873-766
2-Acetyl-3,4-dibromothiophene ≥95%
2-Acetyl-3,4-dibromothiophene
Hazard: Irritant
Formula:
C₆H₄Br₂OS Melting Pt: 78…80 °C |
CAS Number:
57681-57-9 |
Specification Test Results
Melting point | 78-80°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...