3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Bromo-6-cyanoimidazo[1,2-a]pyridine
046017-250MG
046017-1G
101873-758EA
581.54
USD
101873-758
101873-760
3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile ≥95%
3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile
Hazard: Irritant
Formula:
C₈H₄BrN₃ MW: 222.05 g/mol Melting Pt: 212-213°C |
MDL Number:
MFCD06659617 CAS Number: 885950-21-0 |
Specification Test Results
Melting point | 212-213°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...