3-(Dimethylamino)-1-(2-pyridinyl)-2-buten-1-one ≥95%
Supplier: Matrix Scientific
046015-500MG
046015-1G
046015-5G
101873-748EA
375.38
USD
101873-748
101873-750
101873-752
3-(Dimethylamino)-1-(2-pyridinyl)-2-buten-1-one ≥95%
3-(Dimethylamino)-1-(2-pyridinyl)-2-buten-1-one
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O MW: 190.25 g/mol Melting Pt: 61…65 °C |
MDL Number:
MFCD06659616 CAS Number: 112677-15-3 |
Specification Test Results
Melting point | 61-65°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...