4-methoxy-1H-indazol-3-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-4-methoxy-1H-indazole, 4-Methoxy-1H-indazol-3-amine

046012-1G 046012-5G
101873-732EA 96.92 USD
101873-732 101873-734
4-methoxy-1H-indazol-3-amine ≥95%
4-methoxy-1H-indazol-3-amine

Hazard: Irritant

Formula: C₈H₉N₃O
MW: 163.18 g/mol
Melting Pt: 85…87 °C
MDL Number: MFCD08276946
CAS Number: 886362-07-8

Order Now

Specification Test Results

Melting point 85-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR