Methyl-4-(3-formyl-2-pyridinyl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(3-formylpyridin-2-yl)benzoate

046008-500MG 046008-1G 046008-5G
101873-708EA 747.69 USD
101873-708 101873-710 101873-712
Methyl-4-(3-formyl-2-pyridinyl)benzenecarboxylate ≥95%
Methyl-4-(3-formyl-2-pyridinyl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₁NO₃
MW: 241.25 g/mol
Melting Pt: 117…119 °C
MDL Number: MFCD06659611
CAS Number: 885950-15-2

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR