2-(3-Thienyl)nicotinaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Thiophen-3-yl)pyridine-3-carbaldehyde

046007-500MG 046007-1G
101873-704EA 653.85 USD
101873-704 101873-706
2-(3-Thienyl)nicotinaldehyde ≥95%
2-(3-Thienyl)nicotinaldehyde

Hazard: Irritant

Formula: C₁₀H₇NOS
MW: 189.24 g/mol
Melting Pt: 60…62 °C
MDL Number: MFCD06659610
CAS Number: 885950-13-0

Order Now

Specification Test Results

Melting point 60-62°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR