6-(3-Thienyl)nicotinaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 6-(Thiophen-3-yl)pyridine-3-carbaldehyde

046006-500MG 046006-1G 046006-5G
101873-698EA 747.69 USD
101873-698 101873-700 101873-702
6-(3-Thienyl)nicotinaldehyde ≥95%
6-(3-Thienyl)nicotinaldehyde

Hazard: Irritant

Formula: C₁₀H₇NOS
MW: 189.24 g/mol
Melting Pt: 97…100 °C
MDL Number: MFCD04115422
CAS Number: 834884-60-5

Order Now

Specification Test Results

Melting point 97-100°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR