6-(2-Amino-1,3-thiazol-4-yl)-2H-1,4-benzoxazin-3(4H)-one hydrochloride ≥95%

Supplier: Matrix Scientific

046001-500MG 046001-1G 046001-5G
101873-668EA 375.38 USD
101873-668 101873-670 101873-672
6-(2-Amino-1,3-thiazol-4-yl)-2H-1,4-benzoxazin-3(4H)-one hydrochloride ≥95%
6-(2-Amino-1,3-thiazol-4-yl)-2H-1,4-benzoxazin-3(4H)-one hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃O₂S
Melting Pt: 304…306 °C
MDL Number: MFCD06659606
CAS Number: 1049742-69-9

Order Now

Specification Test Results

Melting point 304-306°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR