Ethyl-8-methyl-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

045999-500MG 045999-1G
101873-658EA 375.38 USD
101873-658 101873-660
Ethyl-8-methyl-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate ≥95%
Ethyl 8-methyl-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁F₃N₂O₂
Melting Pt: 80-82°C
MDL Number: MFCD06496249
CAS Number: 874830-63-4

Order Now

Specification Test Results

Melting point 80-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR