Imidazo[1,2-a]pyrimidine ≥95%

Supplier: Matrix Scientific

045994-500MG 045994-5G
101873-634EA 278.46 USD
101873-634 101873-638
Imidazo[1,2-a]pyrimidine ≥95%
Imidazo[1,2-a]pyrimidine

Hazard: Irritant

Formula: C₆H₅N₃
MW: 119.1200 g/mol
Melting Pt: 127…129 °C
MDL Number: MFCD06245377
CAS Number: 274-95-3

Order Now

Specification Test Results

Melting point 127-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR