2-Chloro-4-(1H-pyrrol-1-yl)benzenol ≥95%
Supplier: Matrix Scientific
045987-500MG
045987-1G
045987-5G
101873-608EA
921.54
USD
101873-608
101873-610
101873-612
2-Chloro-4-(1H-pyrrol-1-yl)benzenol ≥95%
2-Chloro-4-(1H-pyrrol-1-yl)benzenol
Hazard: Irritant
Formula:
C₁₀H₈ClNO Melting Pt: 96…99 °C |
MDL Number:
MFCD03844861 CAS Number: 886360-85-6 |
Specification Test Results
Melting point | 96-99°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...