2-Chloro-4-(2,5-dimethyl-1H-pyrrol-1-yl)benzenol ≥95%
Supplier: Matrix Scientific
045986-500MG
045986-1G
045986-5G
101873-602EA
921.54
USD
101873-602
101873-604
101873-606
2-Chloro-4-(2,5-dimethyl-1H-pyrrol-1-yl)benzenol ≥95%
2-Chloro-4-(2,5-dimethyl-1H-pyrrol-1-yl)benzenol
Hazard: Irritant
Formula:
C₁₂H₁₂ClNO Melting Pt: 89…91 °C |
MDL Number:
MFCD03844860 CAS Number: 647841-63-2 |
Specification Test Results
Melting point | 89-91°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...