2-{[(4-Fluorophenyl)sulfonyl]amino}acetic acid ≥95%

Supplier: Matrix Scientific

045985-500MG 045985-1G 045985-5G
101873-596EA 181.54 USD
101873-596 101873-598 101873-600
2-{[(4-Fluorophenyl)sulfonyl]amino}acetic acid ≥95%
2-{[(4-Fluorophenyl)sulfonyl]amino}acetic acid

Hazard: Irritant

Formula: C₈H₈FNO₄S
Melting Pt: 158…160 °C
MDL Number: MFCD02091617
CAS Number: 13029-71-5

Order Now

Specification Test Results

Melting point 158-160°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR