Methyl-5-amino-2-morpholinobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

045979-500MG 045979-1G 045979-5G
101873-572EA 375.38 USD
101873-572 101873-574 101873-576
Methyl-5-amino-2-morpholinobenzenecarboxylate ≥95%
Methyl-5-amino-2-morpholinobenzenecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₃
Melting Pt: 118…120 °C
MDL Number: MFCD00277666
CAS Number: 83909-35-7

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR