2,6-Dimethyl-4-(4-nitrophenyl)morpholine ≥95%
Supplier: Matrix Scientific
045978-500MG
045978-5G
101873-566EA
181.54
USD
101873-566
101873-570
2,6-Dimethyl-4-(4-nitrophenyl)morpholine ≥95%
2,6-Dimethyl-4-(4-nitrophenyl)morpholine
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₃ Melting Pt: 113…116 °C |
MDL Number:
MFCD00105780 CAS Number: 29842-64-6 |
Specification Test Results
Melting point | 113-116°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...