Methyl-2-morpholino-5-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

045977-500MG 045977-1G 045977-5G
101873-560EA 278.46 USD
101873-560 101873-562 101873-564
Methyl-2-morpholino-5-nitrobenzenecarboxylate ≥95%
Methyl-2-morpholino-5-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₅
Melting Pt: 113…116 °C
MDL Number: MFCD00277668
CAS Number: 83909-55-1

Order Now

Specification Test Results

Melting point 113-116°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR