1-(3-Aminophenyl)-2-(3,5-dimethyl-1H-pyrazol-1-yl)-1-ethanone ≥95%
Supplier: Matrix Scientific
045976-500MG
045976-1G
045976-5G
101873-554EA
278.46
USD
101873-554
101873-556
101873-558
1-(3-Aminophenyl)-2-(3,5-dimethyl-1H-pyrazol-1-yl)-1-ethanone ≥95%
1-(3-Aminophenyl)-2-(3,5-dimethyl-1H-pyrazol-1-yl)-1-ethanone
Hazard: Irritant
Formula:
C₁₃H₁₅N₃O Melting Pt: 140…143 °C |
MDL Number:
MFCD03617767 CAS Number: 956206-44-3 |
Specification Test Results
Melting point | 140-143°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...