Ethyl-1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxylate ≥95%
Supplier: Matrix Scientific
045972-500MG
045972-1G
045972-5G
101873-530EA
181.54
USD
101873-530
101873-532
101873-534
Ethyl-1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxylate
Hazard: Irritant
Formula:
C₁₂H₁₆ClN₃O₂ MW: 269.73 g/mol Melting Pt: 55…60 °C |
MDL Number:
MFCD03425828 CAS Number: 252263-47-1 |
Specification Test Results
Melting point | 55-60°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...