{1-[2-(Aminomethyl)phenyl]-4-piperidinyl}methanol ≥95%

Supplier: Matrix Scientific

045969-500MG 045969-1G 045969-5G
101873-512EA 384.62 USD
101873-512 101873-514 101873-516
{1-[2-(Aminomethyl)phenyl]-4-piperidinyl}methanol ≥95%
{1-[2-(Aminomethyl)phenyl]-4-piperidinyl}methanol

Hazard: Irritant

Formula: C₁₃H₂₀N₂O
MW: 220.32 g/mol
Melting Pt: 96…99 °C
MDL Number: MFCD03425826
CAS Number: 220060-79-7

Order Now

Specification Test Results

Melting point 96-99°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR