4-Amino-4,5-dihydro-6H-cyclopenta[b]thiophen-6-one hydrochloride ≥95%

Supplier: Matrix Scientific

045965-500MG 045965-1G
101873-496EA 921.54 USD
101873-496 101873-498
4-Amino-4,5-dihydro-6H-cyclopenta[b]thiophen-6-one hydrochloride ≥95%
4-Amino-4,5-dihydro-6H-cyclopenta[b]thiophen-6-one hydrochloride

Hazard: Irritant

Formula: C₇H₈ClNOS
Melting Pt: 225 °C
MDL Number: MFCD03791250
CAS Number: 108046-16-8

Order Now

Specification Test Results

Melting point 225°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR