5-(2-Thienyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Thiophen-2-yl)-1H-pyrazole

045960-500MG 045960-1G 045960-5G
101873-468EA 747.69 USD
101873-468 101873-470 101873-472
5-(2-Thienyl)-1H-pyrazole ≥95%
5-(2-Thienyl)-1H-pyrazole

Hazard: Irritant

Formula: C₇H₆N₂S
MW: 150.2 g/mol
Melting Pt: 97…99 °C
MDL Number: MFCD00059740
CAS Number: 19933-24-5

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR