Dimethyl-2-(5-nitro-2-pyridinyl)malonate ≥95%
Supplier: Matrix Scientific
Synonyms:
Dimethyl 2-(5-nitropyridin-2-yl)malonate
045959-500MG
045959-1G
045959-5G
101873-462EA
93.85
USD
101873-462
101873-464
101873-466
Dimethyl-2-(5-nitro-2-pyridinyl)malonate ≥95%
Dimethyl-2-(5-nitro-2-pyridinyl)malonate
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O₆ MW: 254.2 g/mol Melting Pt: 103.5 °C |
MDL Number:
MFCD03425824 CAS Number: 68719-87-9 |
Specification Test Results
Melting point | 103.5°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...