3-{[(Benzyloxy)carbonyl]amino}-3-(4-methylphenyl)propanoic acid ≥95%

Supplier: Matrix Scientific

045956-500MG 045956-1G
101873-452EA 556.92 USD
101873-452 101873-454
3-{[(Benzyloxy)carbonyl]amino}-3-(4-methylphenyl)propanoic acid ≥95%
3-{[(Benzyloxy)carbonyl]amino}-3-(4-methylphenyl)propanoic acid

Hazard: Irritant

Formula: C₁₈H₁₉NO₄
Melting Pt: 163…165 °C
MDL Number: MFCD03001314
CAS Number: 406726-23-6

Order Now

Specification Test Results

Melting point 163-165°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR