3-{[(Benzyloxy)carbonyl]amino}-3-(2-thienyl)propanoic acid ≥95%

Supplier: Matrix Scientific

045954-500MG 045954-1G
101873-448EA 556.92 USD
101873-448 101873-450
3-{[(Benzyloxy)carbonyl]amino}-3-(2-thienyl)propanoic acid ≥95%
3-{[(Benzyloxy)carbonyl]amino}-3-(2-thienyl)propanoic acid

Hazard: Irritant

Formula: C₁₅H₁₅NO₄S
Melting Pt: 117…119 °C
MDL Number: MFCD03001313
CAS Number: 73457-03-1

Order Now

Specification Test Results

Melting point 117-119°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR