4-Fluoro-7-nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

045953-500MG 045953-1G 045953-5G
101873-442EA 921.54 USD
101873-442 101873-444 101873-446
4-Fluoro-7-nitro-1H-indazole ≥95%
4-Fluoro-7-nitro-1H-indazole

Hazard: Irritant

Formula: C₇H₄FN₃O₂
MW: 181.13 g/mol
Melting Pt: 190…192 °C
MDL Number: MFCD03787365
CAS Number: 866144-02-7

Order Now

Specification Test Results

Melting point 190-192°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR