3-[(tert-Butoxycarbonyl)amino]-3-(3-hydroxyphenyl)propanoic acid ≥95%

Supplier: Matrix Scientific

045951-500MG 045951-1G
101873-432EA 556.92 USD
101873-432 101873-434
3-[(tert-Butoxycarbonyl)amino]-3-(3-hydroxyphenyl)propanoic acid ≥95%
3-[(tert-Butoxycarbonyl)amino]-3-(3-hydroxyphenyl)propanoic acid

Hazard: Irritant

Formula: C₁₄H₁₉NO₅
Melting Pt: 130…132 °C
MDL Number: MFCD02090711
CAS Number: 284493-68-1

Order Now

Specification Test Results

Melting point 130-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR