N-(3-Fluoro-2-methyl-6-nitrophenyl)acetamide ≥95%

Supplier: Matrix Scientific

045950-500MG 045950-1G 045950-5G
101873-426EA 375.38 USD
101873-426 101873-428 101873-430
N-(3-Fluoro-2-methyl-6-nitrophenyl)acetamide ≥95%
N-(3-Fluoro-2-methyl-6-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₉H₉FN₂O₃
Melting Pt: 145…148 °C
MDL Number: MFCD03787363
CAS Number: 156243-41-3

Order Now

Specification Test Results

Melting point 145-148°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR