3-Amino-3-(1-naphthyl)propanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-3-(naphthalen-1-yl)propanoic acid

045949-500MG 045949-1G 045949-5G
101873-420EA 278.46 USD
101873-420 101873-422 101873-424
3-Amino-3-(1-naphthyl)propanoic acid ≥95%
3-Amino-3-(1-naphthyl)propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₃NO₂
MW: 215.25 g/mol
Melting Pt: 217…219 °C
MDL Number: MFCD00269725
CAS Number: 100393-41-7

Order Now

Specification Test Results

Melting point 217-219°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR