N-(3-Fluoro-2-methylphenyl)acetamide ≥95%

Supplier: Matrix Scientific

045948-500MG 045948-1G 045948-5G
101873-414EA 278.46 USD
101873-414 101873-416 101873-418
N-(3-Fluoro-2-methylphenyl)acetamide ≥95%
N-(3-Fluoro-2-methylphenyl)acetamide

Hazard: Irritant

Formula: C₉H₁₀FNO
Melting Pt: 132…133 °C
MDL Number: MFCD03787362
CAS Number: 322-33-8

Order Now

Specification Test Results

Melting point 132-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR