3-(1,3-Benzodioxol-5-yl)-3-[(tert-butoxycarbonyl)amino]propanoic acid ≥95%

Supplier: Matrix Scientific

045941-500MG 045941-1G
101873-376EA 556.92 USD
101873-376 101873-378
3-(1,3-Benzodioxol-5-yl)-3-[(tert-butoxycarbonyl)amino]propanoic acid ≥95%
3-(1,3-Benzodioxol-5-yl)-3-[(tert-butoxycarbonyl)amino]propanoic acid

Hazard: Irritant

Formula: C₁₅H₁₉NO₆
Melting Pt: 158…160 °C
MDL Number: MFCD02090675
CAS Number: 149520-06-9

Order Now

Specification Test Results

Melting point 158-160°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR