3-(2,4-Dichlorophenyl)-1-oxo-3,4-dihydro-1H-isochromene-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

045937-500MG 045937-1G 045937-5G
101873-360EA 278.46 USD
101873-360 101873-362 101873-364
3-(2,4-Dichlorophenyl)-1-oxo-3,4-dihydro-1H-isochromene-4-carboxylic acid ≥95%
3-(2,4-Dichlorophenyl)-1-oxo-3,4-dihydro-1H-isochromene-4-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₀Cl₂O₄
Melting Pt: 216…219 °C
MDL Number: MFCD03001309
CAS Number: 400751-70-4

Order Now

Specification Test Results

Melting point 216-219°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR