N-[4-(Trifluoromethoxy)phenyl]urea ≥95%

Supplier: Matrix Scientific

045935-500MG 045935-1G 045935-5G
101873-348EA 181.54 USD
101873-348 101873-350 101873-352
N-[4-(Trifluoromethoxy)phenyl]urea ≥95%
N-[4-(Trifluoromethoxy)phenyl]urea

Hazard: Irritant

Formula: C₈H₇F₃N₂O₂
MW: 220.15 g/mol
Melting Pt: 153…155 °C
MDL Number: MFCD03788552
CAS Number: 82971-90-2

Order Now

Specification Test Results

Melting point 153-155°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR