1-[4-(2-Propynyloxy)phenyl]-1-ethanone ≥95%
Supplier: Matrix Scientific
045934-500MG
045934-1G
045934-5G
101873-342EA
278.46
USD
101873-342
101873-344
101873-346
1-[4-(2-Propynyloxy)phenyl]-1-ethanone ≥95%
1-[4-(2-Propynyloxy)phenyl]-1-ethanone
Hazard: Irritant
Formula:
C₁₁H₁₀O₂ Melting Pt: 81…83 °C |
MDL Number:
MFCD03001308 CAS Number: 34264-14-7 |
Specification Test Results
Melting point | 81-83°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...