5-Chloro-2-(2-propynyloxy)benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

045931-500MG 045931-1G 045931-5G
101873-324EA 278.46 USD
101873-324 101873-326 101873-328
5-Chloro-2-(2-propynyloxy)benzenecarbaldehyde ≥95%
5-Chloro-2-(2-propynyloxy)benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₀H₇ClO₂
Melting Pt: 74…76 °C
MDL Number: MFCD03001306
CAS Number: 224317-64-0

Order Now

Specification Test Results

Melting point 74-76°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR