3-[2-(2-Propynyloxy)phenyl]acrylic acid ≥95%

Supplier: Matrix Scientific

045930-500MG 045930-1G 045930-5G
101873-318EA 375.38 USD
101873-318 101873-320 101873-322
3-[2-(2-Propynyloxy)phenyl]acrylic acid ≥95%
3-[2-(2-Propynyloxy)phenyl]acrylic acid

Hazard: Irritant

Formula: C₁₂H₁₀O₃
Melting Pt: 141…143 °C
MDL Number: MFCD03001305
CAS Number: 400751-66-8

Order Now

Specification Test Results

Melting point 141-143°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR