4-(2-Phenylacetyl)-1H-pyrrole-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
045928-500MG
045928-5G
101873-306EA
800
USD
101873-306
101873-310
4-(2-Phenylacetyl)-1H-pyrrole-2-carboxylic acid ≥95%
4-(2-Phenylacetyl)-1H-pyrrole-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₁NO₃ Melting Pt: 236…238 °C |
MDL Number:
MFCD02571500 CAS Number: 478249-37-5 |
Specification Test Results
Melting point | 236-238°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...