3-Amino-4-{[3-(trifluoromethyl)phenyl]sulfanyl}benzenecarbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Amino-4-{[3-(trifluoromethyl)phenyl]thio}benzonitrile
045927-500MG
045927-1G
045927-5G
101873-300EA
921.54
USD
101873-300
101873-302
101873-304
3-Amino-4-{[3-(trifluoromethyl)phenyl]sulfanyl}benzenecarbonitrile ≥95%
3-Amino-4-{[3-(trifluoromethyl)phenyl]sulfanyl}benzenecarbonitrile
Hazard: Irritant
Formula:
C₁₄H₉F₃N₂S MW: 294.3 g/mol Melting Pt: 149…151 °C |
MDL Number:
MFCD03001303 CAS Number: 400751-63-5 |
Specification Test Results
Melting point | 149-151°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...