[2-(2-Propynyloxy)phenyl]methanol ≥95%

Supplier: Matrix Scientific

045925-500MG 045925-1G 045925-5G
101873-290EA 400 USD
101873-290 101873-292 101873-294
[2-(2-Propynyloxy)phenyl]methanol ≥95%
[2-(2-Propynyloxy)phenyl]methanol

Hazard: Irritant

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Melting Pt: 38…40 °C
MDL Number: MFCD03001302
CAS Number: 130441-68-8

Order Now

Specification Test Results

Melting point 38-40°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR